Please login to your online account to display your discounted pricing

Aluminum Oxide, Powder, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$455.52 - $455.52

Chemical Identifiers

Chemical Name or Material Aluminum Oxide
CAS 1344-28-1
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula Al2O3
Formula Weight 101.96
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002248

 
avantor performance
053605
Poly Bottle 2.5kg Each for $455.52
Description & Specifications

Specifications

Formula Weight 101.96
Chemical Name or Material Aluminum Oxide
CAS 1344-28-1
Physical Form Powder
Quantity 2.5kg
Specific Gravity 1L = 4.00kg
Purity Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Molecular Formula Al2O3