Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum oxide, polishing compound

Polishing compound for IR crystal windows

Chemical Identifiers

Chemical Name or Material Aluminum oxide
CAS 1344-28-1
Name Note Polishing compound
Formula Weight 101.96
MDL Number MFCD00003424
Merck Index 14,356
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA39807LA

  SDS
alfa aesar™
39807LA
57g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 101.96
Quantity 57g
Melting Point 2045°C
RTECS Number BD1200000
Physical Form Particles
MDL Number MFCD00003424
Chemical Name or Material Aluminum oxide
Boiling Point 2980°C
Merck Index 14,356
Particle Size 0.3 Micron
Odor Odorless
CAS 1344-28-1
Name Note Polishing compound
TSCA TSCA
Refractive Index 1.765