missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum oxide, Off-white powder, NanoDur™, Alfa Aesar™

Colloidal dispersion with dispersant

Chemical Identifiers

CAS 1344-28-1
Physical Form Powder
Molecular Formula Al2O3
Formula Weight 101.96
MDL Number MFCD00003424
Chemical Name or Material Aluminum oxide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4492522 SDS Alfa Aesar
4492522
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA4492536 SDS Alfa Aesar
4492536
500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1344-28-1
Al2O3
MFCD00003424
Powder
101.96
Aluminum oxide

Specifications

Powder
100g
Al2O3
MFCD00003424
Aluminum oxide
101.96
Off-white
1344-28-1
Odorless