Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum oxide, gamma-phase, 96+% incl. 3% C

Chemical Identifiers

Chemical Name or Material Aluminum oxide
CAS 1344-28-1
Name Note gamma-phase
Formula Weight 101.96
MDL Number MFCD00003424
Merck Index 14,356
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1286714

  SDS
alfa aesar™
1286714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1286722

  SDS
alfa aesar™
1286722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1286736

  SDS
alfa aesar™
1286736
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 101.96
Chemical Name or Material Aluminum oxide
TSCA TSCA
Odor Odorless
Name Note gamma-phase
Particle Size 3.2mm
Physical Form Tablets
Assay ≥96%
Surface Area 175m2/g
MDL Number MFCD00003424
CAS 1344-28-1
Merck Index 14,356
Quantity 25g
Concentration incl. 3% C
Density 3.5g/mL to 3.9g/mL