missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Aluminum oxide, fused, insulating powder, 99.7+%, Thermo Scientific Chemicals
Used in Dry Filling For thermal Insulation of Electrical Furnaces
$152.01 - $475.63
Chemical Identifiers
CAS | 1344-28-1 |
---|---|
Molecular Formula | Al2O3 |
Molecular Weight (g/mol) | 101.96 |
MDL Number | MFCD00003424 |
InChI Key | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
Synonym | aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum |
PubChem CID | 9989226 |
SMILES | [O--].[O--].[O--].[Al+3].[Al+3] |
Description
Used in dry filling for thermal insulation of electrical furnaces.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Form: approx. 180 micronChemical Identifiers
1344-28-1 | |
101.96 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
9989226 |
Al2O3 | |
MFCD00003424 | |
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
[O--].[O--].[O--].[Al+3].[Al+3] |
Specifications
White | |
1 kg | |
Al2O3 | |
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
[O--].[O--].[O--].[Al+3].[Al+3] | |
101.96 | |
Odorless | |
1.33 hPa (1mmHg) at 2158°C |
1344-28-1 | |
>99.7% | |
MFCD00003424 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
dialuminum;oxygen(2-) | |
9989226 | |
White powder | |
Aluminum oxide |
Safety and Handling
EINECSNumber : 215-691-6
RTECSNumber : BD1200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only