Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum oxide, Drysphere™ desiccant, without indicator

$30.32 - $30.32

Chemical Identifiers

Chemical Name or Material Aluminum oxide, Drysphere™ desiccant
CAS 1344-28-1
Name Note Without indicator
Molecular Formula Al2O3
Linear Formula Al2O3
Formula Weight 101.96
MDL Number MFCD00003424
Merck Index 14,356
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4617222

 
alfa aesar™
4617222
100g Each for $30.32
Description & Specifications

Specifications

Formula Weight 101.96
Chemical Name or Material Aluminum oxide, Drysphere™ desiccant
Odor Odorless
Molecular Formula Al2O3
Linear Formula Al2O3
Merck Index 14,356
Melting Point 2045°C
Quantity 100g
TSCA TSCA
Name Note Without indicator
Particle Size 6.35mm
Physical Form Spheres
Refractive Index 1.765
MDL Number MFCD00003424
CAS 1344-28-1
Boiling Point 2980°C
Color White