missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum oxide desiccant, regular

Manufacturer:  Alfa Aesar™ 44018A3

 View more versions of this product

Catalog No. AA44018A3



Description

Description

  • Dry gases and liquids
  • Gases include: CO2, acetylene, air, Ar, ethylene, He, H2, N2, O2
  • Liquids include: benzene, hexane, oil, pentane
Specifications

Specifications

Aluminum oxide desiccant
1344-28-1
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminum;oxygen(2-)
9989226
Odorless
2kg
Regular
Al2O3
aluminium oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
[O-2].[O-2].[O-2].[Al+3].[Al+3]
101.96
Spheres
3.2mm

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
[O-2].[O-2].[O-2].[Al+3].[Al+3]
Al2O3
MFCD00003424
aluminium oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
dialuminum;oxygen(2-)
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents