missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ Aluminum oxide desiccant, regular
Manufacturer: Alfa Aesar™ 4401836
Description
- Dry gases and liquids
- Gases include: CO2, acetylene, air, Ar, ethylene, He, H2, N2, O2
- Liquids include: benzene, hexane, oil, pentane
Specifications
Aluminum oxide desiccant | |
1344-28-1 | |
MFCD00003424 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
dialuminum;oxygen(2-) | |
9989226 | |
Odorless | |
500g |
Regular | |
Al2O3 | |
aluminium oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
[O-2].[O-2].[O-2].[Al+3].[Al+3] | |
101.96 | |
Spheres | |
3.2mm |
Chemical Identifiers
1344-28-1 | |
101.96 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
9989226 | |
[O-2].[O-2].[O-2].[Al+3].[Al+3] |
Al2O3 | |
MFCD00003424 | |
aluminium oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
dialuminum;oxygen(2-) |
Safety and Handling
TSCA : TSCA