Please login to your online account to display your discounted pricing

Aluminum Oxide, Catalyst Support, Intermediate Surface Area, Med pore diam 1.2 micron, Alfa Aesar

$221.50 - $866.39

Chemical Identifiers

Chemical Name or Material Aluminum Oxide
Name Note Intermediate Surface Area
MDL Number MFCD00003424
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA44693A1

  SDS
alfa aesar
44693A1
1kg Each for $221.50

AA44693A7

  SDS
alfa aesar
44693A7
5kg Each for $866.39
Description & Specifications

Specifications

Quantity 1kg
Odor Odorless
Chemical Name or Material Aluminum Oxide
Particle Size 3.2mm (0.125 in.)
Appearance White pellets
MDL Number MFCD00003424
Name Note Intermediate Surface Area