missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum Oxide, Catalyst Support, Intermediate Surface Area, Med pore diam 1.2 micron, Alfa Aesar

Manufacturer:  Thermo Fisher Scientific 44693A1

 View more versions of this product

Catalog No. AA44693A1



Specifications

Specifications

Aluminum Oxide
1344-28-1
MFCD00003424
abramant
[O-2].[O-2].[O-2].[Al+3].[Al+3]
101.96
Odorless
1kg
Intermediate Surface Area
Al2O3
White pellets
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminum;oxygen(2-)
9989226
3.2mm (0.125 in.)

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
[O-2].[O-2].[O-2].[Al+3].[Al+3]
Al2O3
MFCD00003424
abramant
dialuminum;oxygen(2-)
Documents