missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum Oxide, Catalyst Support, Intermediate Surface Area, Alfa Aesar™

Form: S.A. 12m2/g, contains 17.9% SiO2

Manufacturer:  Alfa Aesar 43856A7

 View more versions of this product

Catalog No. AA43856A7



Specifications

Specifications

Aluminum Oxide
1344-28-1
MFCD00003424
abramant
[O-2].[O-2].[O-2].[Al+3].[Al+3]
101.96
Odorless
White
Intermediate Surface Area
Al2O3
White pellets
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminum;oxygen(2-)
9989226
3.2mm (0.125 in.)
5kg

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
[O-2].[O-2].[O-2].[Al+3].[Al+3]
Al2O3
MFCD00003424
abramant
dialuminum;oxygen(2-)
Documents