Please login to your online account to display your discounted pricing

Aluminum oxide, catalyst support, high surface area, 1/4in. Rings, Alfa Aesar™

S.A. 150 m2/g, medium pore diameter 130Å, total pore volume 0.77cc/g

$235.51 - $922.03

Chemical Identifiers

Chemical Name or Material Aluminum oxide
Name Note catalyst support, high surface area, 1/4in. Rings
MDL Number MFCD00003424
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43858A1

 
alfa aesar
43858A1
1kg Each for $235.51

AA43858A7

 
alfa aesar
43858A7
5kg Each for $922.03
Description & Specifications

Specifications

Quantity 1kg
Chemical Name or Material Aluminum oxide
Name Note catalyst support, high surface area, 1/4in. Rings
MDL Number MFCD00003424