Please login to your online account to display your discounted pricing

Aluminum Oxide, Basic, Brockmann Activity Grade I, Powder, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 2.5kg
Packaging Poly Bottle

Manufacturer: avantor performance  053905

 View more versions of this product

Catalog No. 02002241

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 101.96
Chemical Name or Material Aluminum Oxide, Basic
Molecular Formula Al2O3
CAS 1344-28-1
Physical Form Powder
Packaging Poly Bottle
Purity Grade BAKER ANALYZED™ Reagent
Specific Gravity 1L = 4.00kg
Name Note Brockmann Activity Grade I
Quantity 2.5kg

Specifications

Chemical Name or Material Aluminum Oxide, Basic
CAS 1344-28-1
Name Note Brockmann Activity Grade I
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula Al2O3
Formula Weight 101.96

    For Chromatography