Please login to your online account to display your discounted pricing

Alumina, Calcined Aluminum Oxide, Certified, Fisher Chemical

$162.77 - $372.54

Chemical Identifiers

Chemical Name or Material Aluminum Oxide Anhydrous
CAS 1344-28-1
Purity Grade Certified Grade
Physical Form Solid
Molecular Formula Al2O3
Formula Weight 101.96
MDL Number MFCD00003424
Synonym Alumina
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

A591-3

  SDS
fisher scientific company
FLA5913
Glass Bottle 3kg Each for $372.54
Case of 4 Each for $1,490.16

A591-500

  SDS
fisher scientific company
FLA591500
Glass Bottle 500g Each for $162.77
Case of 6 Each for $976.62
Description & Specifications

Specifications

Synonym Alumina
Melting Point 2000°C
Purity Grade Certified Grade
Boiling Point 2980°C
Formula Weight 101.96
Quantity 3kg
Physical Form Solid
Color White
Sulfate 0.07% max.
Chloride 0.005% max.
Identification Pass Test
Packaging Glass Bottle
Loss on Ignition 1% max.
Chemical Name or Material Aluminum Oxide Anhydrous
CAS 1344-28-1
MDL Number MFCD00003424
Alkalis and Alkaline Earths 0.5% max.
Heavy Metals (as Pb) 0.002% max.
Silicate 0.04% max.
Water_soluble Salts 0.75% max.
Iron (Fe) 0.005% max.
Molecular Formula Al2O3
ChemAlert Storage Symbol Gray

  • Typical surface area 0.4 m2/g
  • Known for its insulative properties and hardness

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling