Learn More
Aluminum oxide, alpha-phase, gamma-phase, 99.99% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01455836
Description
It is used in stainless steel coating and in microwave applications. It is used as an adsorbent, desiccating agent, grinding or blending agent, abrasive agent and catalyst, and in the manufacture of dental cements and refractories. Industrial applications include electrical and electronic products, metal Products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Aluminum oxide | |
alpha-phase, gamma-phase | |
Powder | |
(metals basis) | |
MFCD00003424 | |
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
dialuminium(3+) trioxidandiide | |
9989226 | |
99.99% |
2045°C | |
1344-28-1 | |
500 g | |
Al2O3 | |
14,356 | |
Water insoluble. Soluble in hexane,toluene. | |
[O--].[O--].[O--].[Al+3].[Al+3] | |
101.96 | |
101.96 | |
Odorless |
Chemical Identifiers
1344-28-1 | |
101.96 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
9989226 |
Al2O3 | |
MFCD00003424 | |
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
[O--].[O--].[O--].[Al+3].[Al+3] |
Safety and Handling
EINECSNumber : 215-691-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only