missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Aluminum oxide, alpha-phase, 99.997% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04726114
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2980°C | |
2045°C | |
White | |
25 g | |
Al2O3 | |
14,356 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
dialuminum;oxygen(2-) | |
9989226 | |
99.997% | |
3.965 g/mL |
Aluminum oxide | |
1344-28-1 | |
Crystalline | |
(metals basis) | |
MFCD00003424 | |
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
[O--].[O--].[O--].[Al+3].[Al+3] | |
101.96 | |
101.96 | |
Odorless |
Chemical Identifiers
1344-28-1 | |
101.96 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
9989226 |
Al2O3 | |
MFCD00003424 | |
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
[O--].[O--].[O--].[Al+3].[Al+3] |
Safety and Handling
EINECSNumber : 215-691-6
RTECSNumber : BD1200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only