Please login to your online account to display your discounted pricing

Aluminum oxide, Alfa Aesar™, NanoArc™

30% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion w/ dispersant

Chemical Identifiers

Chemical Name or Material Aluminum oxide
Name Note Sold in collaboration with Nanophase Technologies Corporation
CAS 1344-28-1
Physical Form Dry powder
Molecular Formula Al2O3
Formula Weight 101.96g/mol
MDL Number MFCD00003424
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4540622

 
alfa aesar
45406-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4540636

 
alfa aesar
45406-36
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Appearance Off-white powder
Density 1.2-1.8g/cm3
Color Off-white
Quantity 100g
Chemical Name or Material Aluminum oxide
Name Note Sold in collaboration with Nanophase Technologies Corporation
CAS 1344-28-1
Physical Form Dry powder
Molecular Formula Al2O3
Formula Weight 101.96g/mol
MDL Number MFCD00003424
Health Hazard 1 H226-H319
DOT Information DOT Class: 3, Packaging Group: III
Safety and Handling

H226-H319