missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum oxide, activated, neutral, gamma-phase, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4326622

 View more versions of this product

Catalog No. AA4326622



Specifications

Specifications

Aluminum oxide
1344-28-1
(metals basis)
MFCD00003424
aluminium oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
[O-2].[O-2].[O-2].[Al+3].[Al+3]
101.96
101.96
99.9%
Odorless
100m2/g
100g
Activated, neutral, gamma-phase
99.9% (Metals basis)
Al2O3
14,356
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminum;oxygen(2-)
9989226
Powder
Tap density: 2.4 to 5.0g/cm3
40 Micron APS
White

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
[O-2].[O-2].[O-2].[Al+3].[Al+3]
Al2O3
MFCD00003424
aluminium oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
dialuminum;oxygen(2-)
Safety and Handling

Safety and Handling

RTECSNumber : BD1200000

TSCA : TSCA

Documents