Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum oxide, activated, catalyst support, 99% (metals basis)

Chemical Identifiers

Chemical Name or Material Aluminum oxide
CAS 1344-28-1
Name Note Activated, catalyst support
Formula Weight 101.96g/mol
MDL Number MFCD00003424
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3126830

 
alfa aesar™
31268-30
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA31268A1

 
alfa aesar™
31268-A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Odorless
Particle Size 3.2mm
Surface Area 90m2/g
Melting Point 2045°C
Quantity 250g
Chemical Name or Material Aluminum oxide
Name Note Activated, catalyst support
CAS 1344-28-1
Physical Form Spheres
Assay 99%
Assay Percent Notes (metals basis)
Formula Weight 101.96g/mol
MDL Number MFCD00003424
RTECS Number BD1200000
TSCA TSCA