missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum oxide, 99.99% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer:  Thermo Fisher Scientific 1062736

 View more versions of this product

Catalog No. AA1062736



Specifications

Specifications

Aluminum oxide
99.995% (Metal basis)
MFCD00003424
abramant
[O-2].[O-2].[O-2].[Al+3].[Al+3]
101.96
101.96
Odorless
500g
1344-28-1
Al2O3
White powder
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminum;oxygen(2-)
9989226
Powder
White

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
[O-2].[O-2].[O-2].[Al+3].[Al+3]
Al2O3
MFCD00003424
abramant
dialuminum;oxygen(2-)
Safety and Handling

Safety and Handling

EINECSNumber : 215-691-6

Documents