missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum oxide, 99.5%, Alfa Aesar™, NanoDur™

S. A. 32-40m2/g

Manufacturer:  Alfa Aesar 4493122

 View more versions of this product

Catalog No. AA4493122



Specifications

Specifications

Aluminum oxide
1344-28-1
Al2O3
White powder
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminum;oxygen(2-)
9989226
Powder
Odorless
100g
Sold in collaboration with Nanophase Technologies Corporation
99.5%
MFCD00003424
aluminium oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
[O-2].[O-2].[O-2].[Al+3].[Al+3]
101.96
101.96
3.965g/cm3
White

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
[O-2].[O-2].[O-2].[Al+3].[Al+3]
Al2O3
MFCD00003424
aluminium oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
dialuminum;oxygen(2-)
Safety and Handling

Safety and Handling

EINECSNumber : 215-691-6

SDS Documents