Please login to your online account to display your discounted pricing

Aluminum oxide, 99.5%, Alfa Aesar™, NanoDur™

S. A. 32-40m2/g

$66.30 - $275.74

Chemical Identifiers

Chemical Name or Material Aluminum oxide
Name Note Sold in collaboration with Nanophase Technologies Corporation
CAS 1344-28-1
Physical Form Powder
Assay Percent Range 99.5%
Molecular Formula Al2O3
Formula Weight 101.96
MDL Number MFCD00003424
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4493122

  SDS
alfa aesar
4493122
100g Each for $66.30

AA4493136

  SDS
alfa aesar
4493136*
500g Each for $275.74
Description & Specifications

Specifications

Formula Weight 101.96
Quantity 100g
Color White
EINECS Number 215-691-6
Assay Percent Range 99.5%
Chemical Name or Material Aluminum oxide
Density 3.965g/cm3
Physical Form Powder
MDL Number MFCD00003424
Molecular Formula Al2O3
CAS 1344-28-1
Odor Odorless
Appearance White powder
Name Note Sold in collaboration with Nanophase Technologies Corporation