missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum oxalate hydrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 044149A1

 View more versions of this product

Catalog No. AA44149A1



Description

Description

Aluminum oxalate is used as a mordant for printing textiles and dyeing cotton. It finds use as a laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Aluminum oxalate hydrate
White
Al2(C2O4)3·xH2O
1 kg
aluminum oxalate, dialuminium trioxalate, unii-dq79q766vn, aluminum, tris ethanedioato 2-di, dialuminum oxalate, aluminum oxalate hydrate, dialuminium 3+ trioxalate ion, dialuminium 3+ trioxalate, aluminum oxalate hydrate 10g, dialuminium 3+ ion trioxalate
ZCLVNIZJEKLGFA-UHFFFAOYSA-H
dialuminum;oxalate
93561
99%
814-87-9
C6Al2O12
MFCD00050791
14,355
Insoluble in water.
[Al+3].[Al+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
318.02
318.02 (Anhydrous)
Chunks and/or Powder

Chemical Identifiers

814-87-9
318.02
ZCLVNIZJEKLGFA-UHFFFAOYSA-H
93561
[Al+3].[Al+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
C6Al2O12
MFCD00050791
aluminum oxalate, dialuminium trioxalate, unii-dq79q766vn, aluminum, tris ethanedioato 2-di, dialuminum oxalate, aluminum oxalate hydrate, dialuminium 3+ trioxalate ion, dialuminium 3+ trioxalate, aluminum oxalate hydrate 10g, dialuminium 3+ ion trioxalate
dialuminum;oxalate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c

H302+H312

EINECSNumber : 212-407-2

RTECSNumber : BD0516250

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only