Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum, Oil based standard solution, Specpure™, Al 1000μg/g

Chemical Identifiers

Chemical Name or Material Aluminum Oil based standard solution
Name Note Al 1000μg/g
Purity Grade Specpure™
MDL Number MFCD00134029
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1422918

  SDS
alfa aesar™
1422918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Chemical Name or Material Aluminum Oil based standard solution
Quantity 50g
Name Note Al 1000μg/g
Purity Grade Specpure™
MDL Number MFCD00134029
TSCA TSCA