Please login to your online account to display your discounted pricing

Aluminum nitride Rod, Round; Diameter (mm), 10, Alfa Aesar™

$438.69 - $438.69

Chemical Identifiers

Chemical Name or Material Aluminum nitride Rod
CAS 24304-00-5
Name Note Round; Diameter (mm), 10
MDL Number MFCD00003429
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA42998DM

 
alfa aesar
42998DM
Each for $438.69
Description & Specifications

Specifications

Quantity 100mm
Color White to Pale Yellow
Melting Point >2200°C
Chemical Name or Material Aluminum nitride Rod
Odor Odorless
TSCA TSCA
Name Note Round; Diameter (mm), 10
Physical Form Rod
MDL Number MFCD00003429
CAS 24304-00-5
RTECS Number BD1055000
Density 3.26