Please login to your online account to display your discounted pricing

Aluminum nitride Rod, Round, 22mm diameter, Alfa Aesar™

$1,143.69 - $1,211.42

Chemical Identifiers

Chemical Name or Material Aluminum nitride
CAS 24304-00-5
MDL Number MFCD00003429
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43000DO

 
alfa aesar
43000DO
150mm Each for $1,143.69

AA43000DR

 
alfa aesar
43000DR
200mm Each for $1,211.42
Description & Specifications

Specifications

Quantity 150mm
Color White to pale yellow
Chemical Name or Material Aluminum nitride
Odor Odorless
MDL Number MFCD00003429
Appearance White to pale yellow rod
CAS 24304-00-5