missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum Nitride Paste, Alfa Aesar™

Manufacturer:  Alfa Aesar 4395922

 View more versions of this product

Catalog No. AA4395922



Specifications

Specifications

Aluminium Nitride
24304-00-5
MFCD00003429
aluminum nitride, aluminum nitride, aluminium nitride, unii-7k47d7p3m0, aluminium nitride, aluminum nitride paste, aluminum nitride 50g, aluminium nitride grade a, aluminium nitride grade b, aluminium nitride grade c
N#[Al]
40.989
CHEBI:50884
Gray
Air set in 1-4 hours, heat cure in 2 hours at 200°C
AlN
Gray paste
PIGFYZPCRLYGLF-UHFFFAOYSA-N
azanylidynealumane
90455
Odorless
100g

Chemical Identifiers

24304-00-5
40.989
PIGFYZPCRLYGLF-UHFFFAOYSA-N
90455
azanylidynealumane
AlN
MFCD00003429
aluminum nitride, aluminum nitride, aluminium nitride, unii-7k47d7p3m0, aluminium nitride, aluminum nitride paste, aluminum nitride 50g, aluminium nitride grade a, aluminium nitride grade b, aluminium nitride grade c
CHEBI:50884
N#[Al]
Documents