missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum nitride, N 32.0% min

Manufacturer:  Alfa Aesar™ 11546A1

 View more versions of this product

Catalog No. AA11546A1



Specifications

Specifications

Aluminum nitride
AlN
14,353
PIGFYZPCRLYGLF-UHFFFAOYSA-N
azanylidynealumane
90455
40.99
N 32.0% min.
−325 Mesh
typically <4 Micron APS
White
1kg
24304-00-5
MFCD00003429
aluminium nitride
N#[Al]
40.989
CHEBI:50884
Powder
3.26g/mL
Ammonia-like
Moisture Sensitive
>2200°C

Chemical Identifiers

24304-00-5
40.989
PIGFYZPCRLYGLF-UHFFFAOYSA-N
90455
azanylidynealumane
AlN
MFCD00003429
aluminium nitride
CHEBI:50884
N#[Al]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : BD1055000

TSCA : TSCA

Documents