missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum nitrate nonahydrate, 98%, Alfa Aesar

Manufacturer:  Thermo Fisher Scientific 1236030

 View more versions of this product

Catalog No. AA1236030



Specifications

Specifications

Aluminum nitrate nonahydrate
7784-27-2
AlH18N3O18
White crystalline
Soluble in water,ethanol,glycerol and methanol.
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
375.129
375.13 (212.99 Anhydrous)
Odorless
White
250g
nonahydrate
98%
MFCD00003419
aluminium 3+ nonahydrate trinitronate
SWCIQHXIXUMHKA-UHFFFAOYSA-N
aluminum;trinitrate;nonahydrate
24567
Crystals
Hygroscopic
73.5°C (135° (decomposition))

Chemical Identifiers

7784-27-2
375.129
SWCIQHXIXUMHKA-UHFFFAOYSA-N
24567
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
AlH18N3O18
MFCD00003419
aluminium 3+ nonahydrate trinitronate
aluminum;trinitrate;nonahydrate
Safety and Handling

Safety and Handling

P280-P305+P351+P338-P310

H318

DOTInformation : DOT Class: 5.1, Packaging Group: III

EINECSNumber : 236-751-8

RTECSNumber : BD1050000

TSCA : Yes

storageNote1 : Hygroscopic

Recommended Storage : Ambient temperatures

Documents