missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum nitrate nonahydrate, 98%, Alfa Aesar

Manufacturer:  Alfa Aesar 1236014

 View more versions of this product

Catalog No. AA1236014



Specifications

Specifications

Aluminum nitrate nonahydrate
7784-27-2
AlH18N3O18
White crystalline
SWCIQHXIXUMHKA-UHFFFAOYSA-N
aluminum;trinitrate;nonahydrate
24567
Crystalline
White
nonahydrate
98%
MFCD00003419
aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminium 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
375.129
375.13
Odorless
25g

Chemical Identifiers

7784-27-2
375.129
SWCIQHXIXUMHKA-UHFFFAOYSA-N
24567
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
AlH18N3O18
MFCD00003419
aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminium 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic
aluminum;trinitrate;nonahydrate
Safety and Handling

Safety and Handling

H272-H315-H319-H303

DOTInformation : DOT Class: 5.1, Packaging Group: III

EINECSNumber : 236-751-8

storageNote1 : Hygroscopic

Documents