missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ Aluminum Nitrate Nonahydrate, ≥98%, ACS Reagent, Honeywell™ Fluka™

ACS Reagent,≥98%

$117.93 - $442.13

Chemical Identifiers

CAS 7784-27-2
Molecular Formula AlH18N3O18
Molecular Weight (g/mol) 375.129
MDL Number MFCD00149132
InChI Key SWCIQHXIXUMHKA-UHFFFAOYSA-N
Synonym aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminium 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic
PubChem CID 24567
IUPAC Name aluminum;trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60045463 View Documents Honeywell Fluka™
237973100G
100g Poly Bottle Each for $117.93
 
60045465 View Documents Honeywell Fluka™
237973500G
500g Poly Bottle Each for $280.19
 
60045464 View Documents Honeywell Fluka™
23797325KG
2.5kg Poly Bottle Each for $442.13
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7784-27-2
375.129
SWCIQHXIXUMHKA-UHFFFAOYSA-N
24567
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
AlH18N3O18
MFCD00149132
aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminium 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic
aluminum;trinitrate;nonahydrate

Specifications

7784-27-2
Al(NO3)3 · 9H2O
NONH for all modes of transport
aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminium 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
375.129
375.13g/mol
ACS Reagent
2.5 to 3.5 at 20°C
73°C
Aluminum Nitrate Nonahydrate
AlH18N3O18
MFCD00149132
Crystals
SWCIQHXIXUMHKA-UHFFFAOYSA-N
aluminum;trinitrate;nonahydrate
24567
≥98%
Poly Bottle
White
100g
Safety and Handling

Safety and Handling

P280-P305 + P351 + P338 + P310

H318

EINECSNumber : 236-751-8

RTECSNumber : BD1050000