missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum nitrate hydrate, 99.999% (metals basis excluding Hg), Alfa Aesar™, Puratronic™

Manufacturer:  Thermo Fisher Scientific 1062606

 View more versions of this product

Catalog No. AA1062606



Specifications

Specifications

Aluminum nitrate hydrate
99.999% (Metals basis excluding Hg)
MFCD00003419
al.3no3.h2o, aluminium 3+ hydrate trinitrate, aluminum nitrate hydrate, aluminum nitrate hydrate, puratronic
ZZCONUBOESKGOK-UHFFFAOYSA-N
aluminum;trinitrate;hydrate
22078336
Crystalline
Hygroscopic
73.5°C
25838-59-9
AlH2N3O10
White crystalline
Soluble in water,methanol,ethanol,and ethylene glycol.
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Al+3]
231.009
212.99 (Anhydrous)
Odorless
White
5g

Chemical Identifiers

25838-59-9
231.009
ZZCONUBOESKGOK-UHFFFAOYSA-N
22078336
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Al+3]
AlH2N3O10
MFCD00003419
al.3no3.h2o, aluminium 3+ hydrate trinitrate, aluminum nitrate hydrate, aluminum nitrate hydrate, puratronic
aluminum;trinitrate;hydrate
Safety and Handling

Safety and Handling

P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c

H272-H315-H319

DOTInformation : DOT Class: 5.1, Packaging Group: III

EINECSNumber : 236-751-8

RTECSNumber : BD1050000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

Documents