missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminum nitrate hydrate, 99.999% (metals basis excluding Hg), Alfa Aesar™, Puratronic™

Manufacturer:  Thermo Fisher Scientific 1062614

 View more versions of this product

Catalog No. AA1062614



Specifications

Specifications

Aluminum nitrate hydrate
99.999% (Metals basis excluding Hg)
MFCD00003419
al.3no3.h2o
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Al+3]
231.009
212.99
Odorless
25g
25838-59-9
AlH2N3O10
White crystalline
ZZCONUBOESKGOK-UHFFFAOYSA-N
aluminum;trinitrate;hydrate
22078336
Crystalline
White

Chemical Identifiers

25838-59-9
231.009
ZZCONUBOESKGOK-UHFFFAOYSA-N
22078336
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Al+3]
AlH2N3O10
MFCD00003419
al.3no3.h2o
aluminum;trinitrate;hydrate
Safety and Handling

Safety and Handling

H272-H301-H315-H319

DOTInformation : DOT Class: 5.1, Packaging Group: III

EINECSNumber : 236-751-8

Documents