Please login to your online account to display your discounted pricing

Aluminum Nitrate, 9-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$888.00 - $888.00

Chemical Identifiers

Chemical Name or Material Aluminum Nitrate
CAS 7784-27-2
Name Note 9-Hydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Al(NO3)3·9H2O
Formula Weight 375.13
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002238

 
avantor performance
052805
Poly Bottle 2.5kg Each for $888.00
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 375.13
Chemical Name or Material Aluminum Nitrate
Molecular Formula Al(NO3)3·9H2O
Name Note 9-Hydrate
Packaging Poly Bottle
CAS 7784-27-2
Quantity 2.5kg