Please login to your online account to display your discounted pricing

Aluminum Nitrate, 9-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Poly Bottle

Manufacturer: avantor performance  0528-01

 View all options for this product

Catalog No. 02002239

 

Description & Specifications

Chemical Identifiers

Packaging Poly Bottle
Quantity 500g
Quantity 500g
Chemical Name or Material Aluminum Nitrate
Name Note 9-Hydrate
CAS 7784-27-2
Purity Grade BAKER ANALYZED™ ACS Reagent
Physical Form Crystal
Molecular Formula Al(NO3)3·9H2O
Formula Weight 375.13g/mol

Specifications

Chemical Name or Material Aluminum Nitrate
CAS 7784-27-2
Name Note 9-Hydrate
Purity Grade BAKER ANALYZED™ ACS Reagent
Molecular Formula Al(NO3)3·9H2O
Formula Weight 375.13g/mol