Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum nickel isopropoxide, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Aluminum nickel isopropoxide
CAS 70504-57-3
Formula Weight 584.92
MDL Number MFCD00143593
UN Number UN3181
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4237504

 
alfa aesar™
4237504
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4237509

 
alfa aesar™
4237509
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 584.92
Chemical Name or Material Aluminum nickel isopropoxide
Sensitivity Moisture Sensitive
Physical Form Powder
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00143593
CAS 70504-57-3
Quantity 2g
UN Number UN3181
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H228-H351-H315-H319-H317-H335
Highly flammable liquid and vapor.
Flammable solid.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Dark
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H228-H351-H315-H319-H317-H335
Highly flammable liquid and vapor.
Flammable solid.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.