Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum nickel isopropoxide, 99.9% (metals basis), 10% w/v in isopropanol

Chemical Identifiers

Chemical Name or Material Aluminum nickel isopropoxide
CAS 70504-57-3
Formula Weight 584.92
MDL Number MFCD00143593
UN Number UN1219
Synonym Nickel aluminum isopropoxide; Aluminum nickel isopropoxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA36590AC

 
alfa aesar™
36590AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA36590AE

 
alfa aesar™
36590AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 584.92
Health Hazard 2 GHS H Statement
H225-H351-H315-H319-H317-H336
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause drowsiness or dizziness.
Chemical Name or Material Aluminum nickel isopropoxide
Concentration 10% w/v in isopropanol
Sensitivity Moisture Sensitive
Boiling Point 120°C (0.6mmHg)
Physical Form Liquid
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00143593
CAS 70504-57-3
Quantity 25mL
UN Number UN1219
Synonym Nickel aluminum isopropoxide; Aluminum nickel isopropoxide
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Green
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H351-H315-H319-H317-H336
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause drowsiness or dizziness.