Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum nickel isopropoxide, 99.9% (m.b.), 10% w/v in IPA, packaged under Ar in resealable ChemSeal™ bottles

Chemical Identifiers

Chemical Name or Material Aluminum nickel isopropoxide
CAS 70504-57-3
Formula Weight 584.92
MDL Number MFCD00143593
UN Number UN1219
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42376AC

  SDS
alfa aesar™
42376AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA42376AE

  SDS
alfa aesar™
42376AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 584.92
Chemical Name or Material Aluminum nickel isopropoxide
Concentration 10% w/v in isopropanol
Sensitivity Moisture Sensitive
Boiling Point 120°C (0.6mmHg)
Assay Percent Notes (metals basis)
MDL Number MFCD00143593
Quantity 25mL
UN Number UN1219
Health Hazard 1 Danger
Storage Note 1
  • Packaged under Argon
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
Packaging ChemSeal™ bottles
Physical Form Liquid
Assay 99.9%
CAS 70504-57-3
Health Hazard 2 GHS H Statement
H225-H351-H315-H319-H317-H336
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

  • Packaged under Argon

Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H351-H315-H319-H317-H336
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause drowsiness or dizziness.

  • Packaged under Argon