Alfa Aesar™ Aluminum Manganese tubing, alloy 3003, 7.14mm (0.281in) OD, 6.43mm (0.253in) ID
Manufacturer:  Alfa Aesar™ 45942RN
 View more versions of this product
Catalog No. AA45942RN
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Al:Mn:Cu; 98.6:1.2:0.12 wt %
5x 90cm
MFCD00198174
Aluminum Manganese tubing, Alloy 3003
OD: 7.14mm (0.281 in.), ID: 6.43mm (0.253 in.)
Documents