Alfa Aesar™ Aluminum Magnesium gauze , alloy 5056, 20 mesh woven from 0.23mm (0.009 in.) dia. wire
Manufacturer:  Alfa Aesar™ 46579FL
 View more versions of this product
Catalog No. AA46579FL
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Al:Mg; 95:5 wt%
75 x 75mm
MFCD00214039
Odorless
Aluminum Magnesium gauze, Alloy 5056
20 mesh woven from 0.23mm (0.009 in.) dia. wire
20 Mesh
Wire Cloth
Documents