Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum lanthanum isopropoxide, 99% (metals basis), 7% w/v in isopropanol

Thin films of metal oxides

Chemical Identifiers

Chemical Name or Material Aluminum lanthanum isopropoxide
CAS 33939-97-8
Formula Weight 928.24
MDL Number MFCD00143591
UN Number UN1219
Synonym Lanthanum aluminum isopropoxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41262AC

  SDS
alfa aesar™
41262AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA41262AE

  SDS
alfa aesar™
41262AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Aluminum lanthanum isopropoxide
Concentration 7% w/v in isopropanol
Boiling Point 208°C (0.5mmHg)
CAS 33939-97-8
Quantity 25mL
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H315-H319-H336
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
Sensitivity Moisture Sensitive
Physical Form Liquid
Assay 99%
Assay Percent Notes (metals basis)
Formula Weight 928.24
MDL Number MFCD00143591
UN Number UN1219
Synonym Lanthanum aluminum isopropoxide
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H315-H319-H336
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.