Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum iodide, ultra dry, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Aluminum iodide
CAS 7784-23-8
Name Note Ultra dry
Formula Weight 407.69
MDL Number MFCD00003427
UN Number UN3260
Merck Index 14,346
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4717006

  SDS
alfa aesar™
4717006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4717014

  SDS
alfa aesar™
4717014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 407.69
Packaging Ampule
Color White to brownish-purple
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
TSCA TSCA
Boiling Point 360°C
UN Number UN3260
Health Hazard 1 Danger
Chemical Name or Material Aluminum iodide
Name Note Ultra dry
Assay Percent Notes (metals basis)
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 3.98g/mL
Physical Form Powder
DOT Information Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
Quantity 5g
Merck Index 14,346
Sensitivity Moisture & Light Sensitive
CAS 7784-23-8
Assay 99.999%
Melting Point 191°C
MDL Number MFCD00003427
Storage Note 1
  • Ampouled under argon
Odor Pungent

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

  • Ampouled under argon