Learn More
Aluminum iodide, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03071422
Description
Aluminum iodide is employed as a catalyst to break certain kinds of C-O and N-O bonds. It cleaves aryl ethers and deoxygenates epoxides It can also be used for the manufacture of iodinated organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
360°C | |
191°C | |
Chunks | |
95% | |
MFCD00003427 | |
Soluble in alcohol,ether,carbon disulfide | |
[Al](I)(I)I | |
407.695 | |
407.69 | |
3.98 |
Aluminum iodide | |
7784-23-8 | |
100 g | |
AlI3 | |
aluminum iodide, aluminum iodide, aluminum triiodide, aluminum iii iodide, aluminum iodide anhydrous, aluminum iodide, technical grade, aluminum triiodide, aluminum iodide metals basis, aluminum iodide, anhydrous, powder trace metals basis | |
CECABOMBVQNBEC-UHFFFAOYSA-K | |
triiodoalumane | |
82222 | |
Light and moisture sensitive |
Chemical Identifiers
7784-23-8 | |
407.695 | |
CECABOMBVQNBEC-UHFFFAOYSA-K | |
82222 | |
[Al](I)(I)I |
AlI3 | |
MFCD00003427 | |
aluminum iodide, aluminum iodide, aluminum triiodide, aluminum iii iodide, aluminum iodide anhydrous, aluminum iodide, technical grade, aluminum triiodide, aluminum iodide metals basis, aluminum iodide, anhydrous, powder trace metals basis | |
triiodoalumane |
Safety and Handling
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 232-054-8
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only