missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Aluminum ICP Standard, 1000 ppm Al in 3% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company PAL1KN500
Specifications
Aluminum ICP Standard | |
Natural Poly Bottle | |
93.0,4.17,1.29 | |
Colorless | |
AlH18N3O18 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] | |
375.129 | |
1 mL = 1 mg Al |
7784-27-2,7697-37-2,7784-27-2,7732-18-5 | |
500 mL | |
96.0,4.34,1.34 | |
1,000ppm Al | |
SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
aluminum;trinitrate;nonahydrate | |
24567 |