Please login to your online account to display your discounted pricing

Aluminum Hydroxide Suspension, For Chloride, Certified, LabChem™

$142.06 - $142.06

Chemical Identifiers

Chemical Name or Material Aluminum Hydroxide Suspension
CAS 7732-18-5
CAS Max % 97.94
CAS 21645-51-2
CAS Max % 2.06
Name Note For Chloride
Purity Grade Certified Grade
Linear Formula Al(OH)3
Formula Weight 78
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC108401

 
lab chem
LC108401
500mL Each for $142.06
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 78
Chemical Name or Material Aluminum Hydroxide Suspension
Decomposition Information Aluminium oxides
Quantity 500mL
Name Note For Chloride
Physical Form Liquid
Chloride Passes Test
Linear Formula Al(OH)3
CAS 7732-18-5
CAS Max % 97.94
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Identification Passes Test
CAS 21645-51-2
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Odor Ammonia
Packaging Poly Bottle
CAS Max % 2.06
Storage Information Room Temperature
Color Cloudy
Solubility Soluble in water
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.