Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum hexafluoro-2,4-pentanedionate, 98+%

In MOCVD

$86.95 - $86.95

Chemical Identifiers

Chemical Name or Material Aluminum hexafluoro-2,4-pentanedionate
CAS 15306-18-0
Molecular Formula C15H3AlF18O6
Formula Weight 648.17
MDL Number MFCD00050051
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4332804

  SDS
alfa aesar™
4332804
2g Each for $86.95

AA4332809

  SDS
alfa aesar™
4332809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Aluminum hexafluoro-2,4-pentanedionate
CAS 15306-18-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Crystalline
Assay ≥98%
Molecular Formula C15H3AlF18O6
Formula Weight 648.17
MDL Number MFCD00050051
Sensitivity Moisture Sensitive
Boiling Point 170°C (dec.)
Melting Point 70° to 73°C
Quantity 2g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.