missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum granules, 8-12mm (0.32-0.47 in.), 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 1009330

 View more versions of this product

Catalog No. AA1009330



Specifications

Specifications

Aluminum granules
8 to 12mm (0.32 to 0.47 in.)
(metals basis)
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum
26.982
CHEBI:33629
99.9%
250g
7429-90-5
Al
adom
[Al]
Odorless
5359268
Granules

Chemical Identifiers

7429-90-5
26.982
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
aluminum
Al
MFCD00134029
adom
CHEBI:33629
[Al]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents