Please login to your online account to display your discounted pricing

Aluminum Foil Rolls for Science Kits


Compatible with multiple projects and science kits.

Manufacturer: fisher science education  AL-5406-R

 View more versions of this product

Catalog No. S05146

  • / Each
 

Description & Specifications

Specifications

Length (English) 25 ft.
Product Type Foil
Material Aluminum
For Use With Multiple projects and in science kits
Width (English) 18 in.
Width (Metric) 46cm