missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum foil, 2.0mm (0.08 in.) thick, Puratronic™, 99.998% (metals basis)

Manufacturer:  Alfa Aesar™ 11372HP

 View more versions of this product

Catalog No. AA11372HP



Specifications

Specifications

Aluminum foil
100 x 500mm
7429-90-5
Al
adom
[Al]
Odorless
5359268
Foil
Puratronic™
≈13.5g/50x50mm
2.0mm (0.08 in.) thick, Temper: as rolled
(metals basis)
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum
26.982
CHEBI:33629
99.998%

Chemical Identifiers

7429-90-5
26.982
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
aluminum
Al
MFCD00134029
adom
CHEBI:33629
[Al]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents