missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Aluminum foil, 1.5mm (0.06 in.) thick, annealed, 99% (metals basis), Si and Fe (combined) typically 1%

Manufacturer:  Alfa Aesar™ 44490GH

 View more versions of this product

Catalog No. AA44490GH



Specifications

Specifications

Aluminum foil
100 x 100mm
7429-90-5
(metals basis)
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum
26.982
CHEBI:33629
99%
≈10.125g/50x50mm
1.5mm (0.06 in.) thick, Annealed, Temper: soft
Si and Fe (combined) typically 1%
Al
adom
[Al]
Odorless
5359268
Foil

Chemical Identifiers

7429-90-5
26.982
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
aluminum
Al
MFCD00134029
adom
CHEBI:33629
[Al]
Safety and Handling

Safety and Handling

EINECSNumber : 231-072-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents